Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-eNOS/NOS III (Thr495) Rabbit anti-Bovine, MilliporeSigma™

Catalog No. 04811MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-811-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-811-MI Supplier MilliporeSigma Supplier No. 04811
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

This Anti-phospho-eNOS/NOS III (Thr495) Antibody is validated for use in western blotting for the detection of phospho-eNOS/NOS III (Thr495).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-eNOS/NOS III (Thr495)
Applications Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
Gene Accession No. NM_000603
Host Species Rabbit
Immunogen Peptide containing the sequence KK[pT]FK in which pT corresponds to phosphothreonine at residue 495 of human eNOS/NOS III
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NOS3, cNOS, ECNOS, eNOS, EC-NOS, NOSIII
Target Species Bovine
Content And Storage −20°C for 24 months. Aliquot to avoid freeze/thaw cycles.
Form Supernatant
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.