Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-erbB-2/HER-2 (Tyr1248) Rabbit anti-Human, Rat, AlexaFluor™488, Polyclonal, MilliporeSigma™

Catalog No. 16235MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-235-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-235-MI Supplier MilliporeSigma Supplier No. 16235
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Alexa Fluor 488 is validated for use in flow cytometry, IC for the detection of phospho-erbB-2/HER-2 (Tyr1248).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-erbB-2/HER-2 (Tyr1248)
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Gene Symbols ERBB2; HER2; HER-2; NGL; herstatin; erbB-2; C-erbB-2; NEU; TKR1; p185erbB2; HER-2/neu
Host Species Rabbit
Immunogen Synthetic phosphopeptide, residues 1243-1255, corresponding to a major autophosphorylation (Tyr1248) site of human Erb B2/HER-2 (C-AENPEpYLGLDVPV)
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001005862.1; NM_004448.2
Target Species Human, Rat
Content And Storage 4°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.