Learn More
Phospho-erbB-2/HER-2 (Tyr1248) Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-erbB-2/HER-2 (Tyr1248) |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Double immunoaffinity purified rabbit polyclonal IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH7.4 |
| Gene Symbols | ERBB2; HER2; C-erbB-2; NEU; HER-2/neu; NGL; herstatin; erbB-2; p185erbB2; HER-2; TKR1 |
| Host Species | Rabbit |
| Immunogen | Synthetic phosphopeptide, residues 1243-1255, corresponding to a major auto-phosphorylation (Tyr1248) site of human erbB-2/HER-2 (C-AENPEpYLGLDVPV). |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.