Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Erk1/2 Rabbit anti-Human, Mouse, Rat, PE, Clone: AW39R, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma FCMAB100P
Description
This Milli-Mark™ Phospho-Erk1/2 antibody is validated for use in Flow and western blotting for the detection of the Milli-Mark™ Phospho-Erk1/2 protein.
Specifications
Phospho-Erk1/2 (Thr202/Tyr204 | |
Polyclonal | |
PE | |
Erk1; Erk2; p44-MAPK; p42-MAPK; MAPK2; PRKM2; p41mapk; PRKM1; p44; ERK2; P42MAPK; ERK; p42; MAPK1; ERT1; ERT2; Mitogen-activated protein kinase; MAP Kinase 2; MAP Kinase 1; MAPK | |
Phosphorylated recombinant peptide corresponding to amino acids surrounding the pTEpY motif in the activation loop of human phospho-Erk1/2, in which the Thr and Tyr residues are phosphorylated | |
100 Tests | |
NP_002736.3 | |
2-8°C for up to 6 months. Protect from light. | |
IgG |
Flow Cytometry, Western Blot | |
AW39R | |
Purified Rabbit monoclonal supernatant conjugated to phycoerythrin in buffer containing PBS with 0.05% sodium azide | |
Rabbit | |
Protein A purfied | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction