Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-FAK (Tyr861) Rabbit anti-Chicken, Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07832MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-832-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-832-MI Supplier MilliporeSigma Supplier No. 07832
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect phospho-FAK (Tyr861) using this Anti-phospho-FAK (Tyr861) Antibody validated for use in western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-FAK (Tyr861)
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene Symbols PTK2; pp125FAK; FAK1; FADK; FAK
Host Species Rabbit
Immunogen Peptide corresponding to amino acid region encompassing the human, mouse, rat, chicken, and frog phospho-FAK (Tyr861)
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_005607.3; NM_153831.2
Target Species Chicken, Human, Mouse
Content And Storage −20°C for 24 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.