Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho-Filamin-A (Ser2152) Clone: PS2,

Catalog No. MABN1834MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1834MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1834MI Supplier MilliporeSigma™ Supplier No. MABN1834
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Filamin-A (Ser2152) Clone: PS2 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Western Blotting

Filamin-A (UniProt P21333; also known as ABP-280, Actin binding protein 280, Alpha-filamin, Endothelial actin-binding protein, Filamin-1, FLN-A, Non-muscle filamin) is encoded by the FLNA (also known as ABP, CSBS, CVD1, FLN, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD) gene (Gene ID 2316) in human. Filamin-A (FLN-A) is an actin-binding protein essential for cytoskeletal rearrangement and cell locomotion. Serine 2152 of FLN-A is a PKA phosphorylation target. FLN-A S2152 phosphorylation is essential for FLN-A-mediated recruitment of signaling proteins, adhesion molecules, and the vesicle-trafficking complex to the peripheral actin rim, so as to stabilize the cortical structure and epithelial membrane. Mutations in FLNA and ARFGEF2 genes cause periventricular heterotopia (PVH or PH), a congenital malformation of cortical development characterized by a reduced brain size and ectopic neuronal nodules along the ventricular neuroepithelium as a result of impaired initiation of neuronal migration. Suppression of either Arfgef2 or Mekk4 results in increased cellular FLN-A level and Ser2152 phosphorylation. In addition, Arfgef2 or Mekk4 deletion causes PVH in knockout mice.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Filamin-A (Ser2152)
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone PS2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P21333
Gene Symbols FLNA, FLN, FLN1
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the Filamin repeat 20 of human phospho-Filamin-A (Ser2152).
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001447
Test Specificity Ser2152 of human and murine Filamin-A isoform 1 (P21333-1 and Q8BTM8-1) corresponds to Ser2144 of human Filamin-A isoform 2 (P21333-2). Equivalent site is not present in murine spliced isoform 2.
Target Species Human, Mouse
Content And Storage 2°C to 8°C for one year from date of shipment
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.