Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Flt-1 (Tyr1213) Rabbit anti-Bovine, Goat, Human, Mouse, Rat, Sheep, Polyclonal, MilliporeSigma™

Catalog No. 07758MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-758-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-758-MI Supplier MilliporeSigma Supplier No. 07758
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-Flt-1 (Tyr1213) Antibody detects level of phospho-Flt-1 (Tyr1213) and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Flt-1 (Tyr1213)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoaffinity purified rabbit IgG in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide.
Gene Symbols FLT1; VEGFR1; FLT; FRT; Flt-1; VEGFR-1
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide containing the sequence (...VR[pY]VN...) in which [pY] corresponds to phospho-tyrosine at residue 1213 of human Flt-1. The immunizing sequence is identical in marmoset, has 14/15 identical amino acids in rat, goat, sheep
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002019.2
Target Species Bovine, Goat, Human, Mouse, Rat, Sheep
Content And Storage 2-8°C for 12 months Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.