Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Focal Adhesion Kinase (Tyr397) Mouse anti-Human, Mouse, Clone: 18, MilliporeSigma™

Catalog No. 051140MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-114-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-114-0MI Supplier MilliporeSigma Supplier No. 051140
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-phospho-Focal Adhesion Kinase (Tyr397) Antibody, clone 18 is validated for use in western blotting for the detection of phospho-Focal Adhesion Kinase (Tyr397).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Focal Adhesion Kinase (Tyr397)
Applications Western Blot
Classification Monoclonal
Clone 18
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 in aqueous buffered solution containing 50% glycerol, BSA, and < 0.09% sodium azide.
Gene Accession No. Q05397
Gene Symbols EC 2.7.10.2, FADK, FAK , FAK1 , pp125FAK
Host Species Mouse
Immunogen Generated from human FAK, a.a. 393-404, phosphorylated on Tyr397.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.