Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-FOXO1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 072126
Description
Use Anti-phospho-FOXO1 (Thr24) Antibody (Rabbit Polyclonal Antibody) validated in western blotting to detect phospho-FOXO1 (Thr24) also known as Forkhead box protein O1.
Specifications
phospho-FOXO1 (Thr24) | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |
Western Blot | |
Unconjugated | |
FOXO1; FOXO1A; FKHR | |
KLH-conjugated linear peptide corresponding to human FOXO1 phosphorylated at Thr24. | |
100 μg | |
NP_002006 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction