Learn More
Specifications
Specifications
| Antigen | phospho-GluR2 (Ser880) |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative. |
| Gene Symbols | GRIA2; GluR-B; GLURB; gluR-B; GLUR2; GluR-2; HBGR2; GluR-K2 |
| Host Species | Rabbit |
| Immunogen | Thyroglobulin-conjugated, synthetic peptide (LVYGIE(PS)VKIA) corresponding to amino acids 873-883 of rat GluR2, with a phosphorylated serine at position 880 |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.