Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-GluR4 (S862) Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB2212
Description
Anti-phospho GluR4 (S862) Antibody is an antibody against phospho GluR4 (S862) for use in western blotting.
Specifications
phospho GluR4 (S862) | |
Polyclonal | |
Serum with 0.05% NaN3 | |
Rabbit | |
50 μL | |
Rat | |
Serum |
Western Blot | |
Unconjugated | |
GRIA4; GLURD; GluR-4; GluR-D | |
Synthetic peptide from rat. | |
Primary | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction