Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-GSK3Β (Ser9) Mouse anti-Human, Mouse, Rat, Clone: 2D3, MilliporeSigma™

Catalog No. 05643MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-643-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-643-MI Supplier MilliporeSigma Supplier No. 05643
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-GSK3β (Ser9) Antibody, clone 2D3 is an antibody against phospho-GSK3β (Ser9) for use in EMSA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-GSK3β (Ser9)
Applications Electromobility Shift Assay, Western Blot
Classification Monoclonal
Clone 2D3
Conjugate Unconjugated
Formulation 100μg of thiophilic and size exclusion chromatography purified mouse IgG1κ in 100μL of antibody storage buffer (PBS with PEG, Sucrose, and 0.09% sodium azide) and 50% glycerol.
Gene Accession No. NM_002093
Gene Symbols GSK3B
Host Species Mouse
Immunogen peptide corresponding to amino acids 6-12 (RTT[pS]AE) of human GSK3beta
Purification Method Thiophilic adsorption chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.