Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-HSP27 (Ser82), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA125983
Description
PA1-25983 detects Heat Shock Protein 27 (phospho S82) from human samples. PA1-25983 will not react with endogenous mouse Heat Shock Protein 27 protein phosphorylated on S86 (S86 in mouse is in a region identical to that surrounding S82 in human Heat Shock Protein 27) in extracts of NIH3T3 cells treatePA1-25983 has been successfully used in Western blot procedures.The PA1-25983 immunogen is a synthetic phosphopeptide corresponding to a portion of human Heat Shock Protein 27 containing threonine 232.Store at 4ºC short term. For extended storage, aliquot and store at -20ºC or below. Avoid freeze/thaw cycles.
Specifications
Phospho-HSP27 pSer82 | |
Polyclonal | |
HSPB1 | |
Phospho-HSP 27, Heat shock protein beta-1, HspB1, HSP25, Heat shock 27kDa protein, HSP 27, Stress-responsive protein 27, SRP27, Estrogen-regulated 24kDa protein, 28kDa heat shock protein, HSPB1, HSP27 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3315 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
P04792 | |
HSPB1 | |
Synthetic phospho peptide (Human) | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction