Learn More
Phospho-Histone H1 Mouse anti-Human, Bovine, Mouse, Vertebrates, Clone: 12D11, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-Histone H1 |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 12D11 |
| Conjugate | Unconjugated |
| Formulation | Supplied as 200μL of culture supernatant with 0.05% sodium azide. |
| Gene Accession No. | NM_005319 |
| Gene Symbols | H1, H1.5 , H1F5 , MGC126630, MGC126632, OTTHUMP00000017748 |
| Host Species | Mouse |
| Immunogen | The immunogen was KLH-conjugated bovine nuclear extract. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.