Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H2A.X (Ser139) Mouse anti-Human, Biotin, Clone: JBW301, MilliporeSigma™

Catalog No. 16193MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-193-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-193-MI Supplier MilliporeSigma Supplier No. 16193
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, biotin conjugate is a Mouse Monoclonal Antibody for detection of phospho-Histone H2A.X (Ser139) also known as H2AXS139P, H2AX histone and has been validated in immunocytochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H2A.X (Ser139)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone JBW301
Conjugate Biotin
Formulation Purified biotin conjugated mouse monoclonal IgG1 in buffer containing 70% storage buffer (0.02MPhosphate buffer, pH7.6, 0.25MNaCl, 0.1% sodium azide) and 30% glycerol
Gene Accession No. NM_002105.2
Gene Symbols H2AFX, H2AX, H2a/x, H2A/X, H2A.X
Host Species Mouse
Immunogen KLH-conjugated, synthetic peptide (CKATQA[pS]QEY) corresponding to amino acids 134-142 of human histone H2A.X. The immunizing sequence has 8 identical amino acids in yeast and mouse.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.