Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H2A.X (Ser139) Mouse anti-Human, Mouse, Rat, Clone: JBW301, MilliporeSigma™

Catalog No. 05636IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-636-IMI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-636-IMI Supplier MilliporeSigma Supplier No. 05636I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301 is a highly specific mouse monoclonal antibody, that targets Histone H2A.X and has been tested in western blotting, immunocytochemistry, Chimmunoprecipitation and Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H2A.X (Ser139)
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone JBW301
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP002096
Gene Symbols H2AFX, H2AX, H2A.X
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to human phospho-Histone H2A. X (Ser139).
Purification Method Protein G purified
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.