Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phospho-Histone H2A.X (Ser139) Mouse anti-Vertebrates, Clone: JBW301
GSA_VA

Catalog No. 05636MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-636-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-636-MI Supplier MilliporeSigma™ Supplier No. 05636
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in Chimmunoprecipitation, immunocytochemistry, immunofluoresence, western blotting. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H2A.X (Ser139)
Applications ChIP Assay, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone JBW301
Conjugate Unconjugated
Formulation Immunoaffinity Purified immunoglobulin in 0.1M Tris-Glycine,pH 7.4, 0.15M NaCl, 0.05% sodium azide as a preservative.
Gene Accession No. NP002096
Gene Symbols H2AFX, H2AX, H2a/x, H2A/X, H2A.X
Host Species Mouse
Immunogen peptide (C-KATQA[pS]QEY) corresponding to amino acids 134-142 of human histone H2A.X
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Vertebrates
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.