Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H2A.X (Thr120) Mouse anti-Human, Clone: 11F5.3, MilliporeSigma™

Catalog No. MABE171MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE171MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE171MI Supplier MilliporeSigma Supplier No. MABE171

Mouse Monoclonal Antibody

Anti-phospho-Histone H2A.X (Thr120) Antibody, clone 11F5.3 is a mouse monoclonal antibody for detection of phospho-Histone H2A.X (Thr120) also known as Histone H2A.X, H2A/X and has been validated in western blot, DB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H2A.X (Thr120)
Applications Dot Blot, Western Blot
Classification Monoclonal
Clone 11F5.3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG3κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Np_002096
Gene Symbols H2Afx, H2Ax
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to Histone H2A.X phosphorylated at Thr120.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.