Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phospho-Histone H3 (Ser10) Rabbit anti-Human, Mouse, AlexaFluor™488, Polyclonal

Catalog No. 06570AF488 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-570-AF488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-570-AF488 Supplier MilliporeSigma™ Supplier No. 06570AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-H3 (Ser10), Alexa Fluor™ 488 Conjugate is a purified rabbit polyclonal that detects Histone H3 phosphorylated at serine 10. This Ab is labeled with Alexa Fluor 488 and validated in immunocytochemistry. The unconjugated version of this antibody is highly published and proven in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Histone H3 (Ser10)
Applications Immunocytochemistry
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation Purified rabbit conjugate in buffer containing PBS, 15mg/mL BSA, and 0.1% Sodium Azide.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen Linear peptide corresponding to phospho-Histone H3 (Ser10).
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.