Learn More
Phospho-Histone H3 (Ser10), Alexa Fluor™ 647 Conjugate Rabbit anti-Human, AlexaFluor™647, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho Histone H3 (Ser10) |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.5mg/mL |
| Conjugate | Alexa Fluor 647 |
| Formulation | Purified rabbit conjugated antibody in buffer containing PBS and 15mg/mL BSA with 0.1% Sodium Azide. |
| Gene Symbols | HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886 |
| Host Species | Rabbit |
| Immunogen | Linear peptide corresponding to Histone H3 phosphorylated at Ser10. |
| Purification Method | Protein A purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.