Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser10) Mouse anti-Human, AlexaFluor™647, Clone: 3H10, MilliporeSigma™

Catalog No. 05806AF647 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-806-AF647 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-806-AF647 Supplier MilliporeSigma Supplier No. 05806AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-phospho Histone H3 (Ser10) antibody, clone 3H10, Alexa Fluor™ 647 is a purified mouse monoclonal for detection of Histone H3 phosphorylated at serine 10.This mAb is fluorescently labeled and validated for use in immunocytochemistry. The unconjugated version has been proven in flow cytometry, immunocytochemistry, immunofluoresence, PIA, western blot, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Histone H3 (Ser10)
Applications Immunocytochemistry
Classification Monoclonal
Clone 3H10, Alexa Fluor™ 647, Alexa Fluor™ 647
Conjugate Alexa Fluor 647
Formulation Purified mouse conjugated antibody in buffer containing PBS and 15mg/mL BSA with 0.1% Sodium Azide.
Gene Accession No. NM_003493
Gene Symbols HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886
Host Species Mouse
Immunogen Linear peptide corresponding to Histone H3 phosphorylated at Ser10.
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.