Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser10) Mouse anti-Human, Clone: 3H10, MilliporeSigma™

Catalog No. 05806MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-806-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-806-MI Supplier MilliporeSigma Supplier No. 05806
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10 is a mouse monoclonal antibody for detection of Histone H3 phosphorylated at serine 10. This purified mAb, also known as H3S10p, is published in peer reviewed journals and has been validated in flow cytometry, immunocytochemistry, immunofluoresence, PIA, western blot, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H3 (Ser10)
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 3H10
Conjugate Unconjugated
Formulation Purified mouse IgG1k in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl and 0.05% sodium azide.
Gene Accession No. NM_003493
Gene Symbols HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886
Host Species Mouse
Immunogen A proprietary immunogen based on a peptide sequence containing phospho-serine corresponding to residue 10 of human histone H3.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.