Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser10) Rat anti-Human, Clone: 6G8B7, MilliporeSigma™

Catalog No. MABE939MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE939MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE939MI Supplier MilliporeSigma Supplier No. MABE939
Add to Cart
Add to Cart

Rat Monoclonal Antibody

This Anti-phospho Histone H3 (Ser10), clone 6G8B7 Antibody is validated for use in Western Blotting and Immunocytochemistry and ELISA for the detection of phospho Histone H3 (Ser10).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho Histone H3 (Ser10)
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 6G8B7
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_002098
Gene Symbols H3F3A, H3.3A, H3F3
Host Species Rat
Immunogen KLH-conjugated linear peptide corresponding to human Histone H3 (Ser10).
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.