Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser10) Mouse anti-Human, Vertebrates, Clone: CMA312, MilliporeSigma™

Catalog No. 051336MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-133-6MI 100 μg
05-133-6SMI 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-133-6MI Supplier MilliporeSigma Supplier No. 051336
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Histone H3 (Ser10) Antibody, clone CMA312 is a mouse monoclonal antibody for detection of phospho-Histone H3 (Ser10) also known as H3S10P, Histone H3 (phospho S10) and has been validated in western blot, ELISA, immunocytochemistry, Multiplexing and Chimmunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H3 (Ser10)
Applications ChIP Assay, ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone CMA312
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in PBS with 0.05% sodium azide.
Gene Accession No. NP_003484.1
Gene Symbols H3.3A, H3.3B , H3F3, MGC87782, MGC87783, OTTHUMP00000035618, OTTHUMP00000035619, OTTHUMP00000035621
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 1-19 of human Histone H3, phosphorylated on Ser10, conjugated to KLH.
Purification Method Protein G chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Vertebrates
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.