Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser10) Mouse anti-Human, Vertebrates, Nematode, Clone: RR002, MilliporeSigma™

Catalog No. 05598MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-598-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-598-MI Supplier MilliporeSigma Supplier No. 05598
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Histone H3 (Ser10) Antibody, clone RR002 is a Mouse Monoclonal Antibody for detection of Histone H3 phosphorylated at serine 10. This purified mAb, also known as Anti-H3S10p, is published in peer reviewed journals and is validated in immunocytochemistry, immunohistochemistry, western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H3 (Ser10)
Applications Immunocytochemistry, Immunohistochemistry
Classification Monoclonal
Clone RR002
Conjugate Unconjugated
Formulation Purified mouse IgG in buffer containing 0.1M Tris Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Gene Accession No. NM_003493
Gene Symbols HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886
Host Species Mouse
Immunogen Peptide (ARK[pS]TGGKAPRKQL-C) corresponding to amino acids 7-20 of human histone H3 with a c-terminal cyteine added for conjugation purposes.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Nematode, Vertebrates
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.