Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Histone H3 (Ser10) Mouse anti-Human, Cy5, Clone: 3H11, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 16218
Description
Anti-Phospho-Histone H3 (Ser10) Antibody, clone 3H10, Cy5 Conjugate is a Mouse Monoclonal Antibody for detection of Histone H3 phosphorylated at serine 10. This purified mAb, also known as H3S10p,is labeled with Cy5.5 NHS ester (Cy5) and validated in immunofluoresence, western blotting.
Specifications
Phospho-Histone H3 (Ser10) | |
Monoclonal | |
Cyanine5 | |
HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886 | |
A proprietary immunogen based on a peptide sequence containing phospho-serine corresponding to residue 10 of human histone H3, clone 3H10. This is a Cy5 conjugate of Anti-phospho-Histone H3 (Ser10), clone 3H10, Catalog # 05-806. | |
100 μg | |
Human | |
Purified |
Immunofluorescence, Western Blot | |
3H11 | |
NM_003493.2 | |
Mouse | |
Protein G purfied | |
Primary | |
4°C for one year | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction