Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser10) Mouse anti-Human, FITC, Clone: 3H10, MilliporeSigma™

Catalog No. 16222MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-222-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-222-MI Supplier MilliporeSigma Supplier No. 16222
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Phospho-Histone H3 (Ser10) Antibody, clone 3H10, FITC Conjugate is a Mouse Monoclonal for detection of Histone H3 phosphorylated at serine 10. This mAb, also known as H3S10p,is labeled with Fluorescein isothiocyanate (FITC), published in peer reviewed journals and validated in western blot, immunocytochemistry and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Histone H3 (Ser10)
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3H10
Conjugate FITC
Formulation Purified mouse monoclonal IgG in buffer containing PBS with 0.05% sodium azide, pH7.1.
Gene Accession No. NP_003484
Gene Symbols HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886
Host Species Mouse
Immunogen A proprietary immunogen based on a peptide sequence containing phospho-serine corresponding to residue 10 of human histone H3. Clone 3H10.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.