Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Ser28) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07145MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-145-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-145-MI Supplier MilliporeSigma Supplier No. 07145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Use Anti-phospho-Histone H3 (Ser28) Antibody (Rabbit Polyclonal Antibody) validated in western blot, immunocytochemistry, DB, PIA, Chimmunoprecipitation to detect phospho-Histone H3 (Ser28) also known as H3F3, H3F3B, H3S28P.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H3 (Ser28)
Applications ChIP Assay, Dot Blot, Immunocytochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols HIST3H3; H3FT
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to Histone H3 phosphorylated at Ser28.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003484.1
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.