Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Histone H3 (Thr11) Rabbit anti-Human, Clone: MC83, MilliporeSigma™

Catalog No. 04789MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-789-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-789-MI Supplier MilliporeSigma Supplier No. 04789
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Anti-phospho-Histone H3 (Thr11) Antibody, clone MC83 is a rabbit monoclonal antibody for detection of phospho-Histone H3 (Thr11) also known as H3T11P, Histone H3 (phospho T11) and has been validated in western blot, PIA, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Histone H3 (Thr11)
Applications Multiplex, Inhibition Assays, Western Blot
Classification Monoclonal
Clone MC83
Conjugate Unconjugated
Formulation Cultured supernantant in 0.05% sodium azide
Gene Accession No. NM_003493
Host Species Rabbit
Immunogen Peptide containing ...KS[pT]G..., in which pT corresponds to phospho-threonine at position 11 of human histone H3
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) H3/t, H3.4, H3/g, H3T, MGC126888, H3t, MGC126886, H3FT, HIST3H3
Target Species Human
Content And Storage −20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.