Learn More
Phospho-Histone H3 (Tyr41) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE1413
Description
This Anti-phospho Histone H3 (Tyr41) Antibody is validated for use in Western Blotting and Dot Blot and Immunohistochemistry for the detection of phospho Histone H3 (Tyr41).
Specifications
phospho Histone H3 (Tyr41) | |
Polyclonal | |
Unconjugated | |
HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ | |
KLH-conjugated linear peptide corresponding to human phospho Histone H3 (Tyr41) near the N-terminus. | |
100 μg | |
NP_003520 | |
2-8°C for 12 months |
Dot Blot, Immunohistochemistry, Western Blot | |
1mg/mL | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human |