Learn More
Phospho-HSP27 (pS82) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho HSP27 (pS82) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol with 1.0mg/mL BSA (IgG, protease free) as a carrier. 0.05% sodium azide. |
| Gene Symbols | HSPB1; HspB1; HS.76067; HMN2B; Hs.76067; HSP27; Hsp25; HSP28; CMT2F; DKFZp586P1322; SRP27 |
| Host Species | Rabbit |
| Immunogen | The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human HSP27 that contains serine 82. |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.