Learn More
Phospho-HSP27 (Ser78) Mouse anti-Human, Clone: JBW502, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-HSP27 (Ser78) |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | JBW502 |
| Conjugate | Unconjugated |
| Formulation | 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% |
| Gene Accession No. | NM_001540 |
| Gene Symbols | HSPB1, Hs.76067, HSP27, Hsp25, HspB1, HSP28, CMT2F, HS.76067, DKFZp586P1322, HMN2B, SRP27 |
| Host Species | Mouse |
| Immunogen | peptide corresponding to amino acids 73-83 of human HSP27 with a C-terminus cysteine added for conjugation (YSRAL[pS]RQLSS-C) |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.