Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-IGF-1R (Tyr1161/Tyr1165/Tyr1166) Rabbit anti-Human, Rat, Mouse, Pig, Xenopus, Chicken, Polyclonal, MilliporeSigma™

Catalog No. ABE332MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE332MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE332MI Supplier MilliporeSigma Supplier No. ABE332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-IGF-1R (Tyr1161/Tyr1165/Tyr1166) Antibody is an antibody against phospho-IGF-1R (Tyr1161/Tyr1165/Tyr1166) for use in western blot, Cell Function Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-IGF-1R (Tyr1161/Tyr1165/Tyr1166)
Applications Functional Assay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols IGF1R
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human IGF-1R phosphorylated at Tyr1161/Tyr1165/Tyr1166.
Purification Method Affinity Purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_000866
Target Species Chicken, Human, Mouse, Pig, Rat, Xenopus species
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.