Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Insulin Receptor (Tyr 1150/1151) Mouse anti-Human, Clone: 10C3, MilliporeSigma™

Catalog No. 04299MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-299-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-299-MI Supplier MilliporeSigma Supplier No. 04299
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect phospho-Insulin Receptor (Tyr 1150/1151) using this Anti-phospho-Insulin Receptor (Tyr 1150/1151) Antibody, clone 10C3 validated for use in EA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Insulin Receptor (Tyr 1150/1151)
Applications Western Blot
Classification Monoclonal
Clone 10C3
Conjugate Unconjugated
Formulation In 1 mL of PBS containing 0.09% sodium azide, PEG, and sucrose and 50% glycerol.
Gene Accession No. NM_000208.2; NM_001079817.1
Gene Symbols INSR, CD220, IR, HHF5
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Tyrosine 1150/1151 on phosphorylated insulin receptor.
Purification Method Thiophilic adsoption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.