Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Insulin Receptor (Tyr 1322) Mouse anti-Canine, Human, Mouse, Clone: 21g12, MilliporeSigma™

Catalog No. 04300MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-300-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-300-MI Supplier MilliporeSigma Supplier No. 04300
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-phospho-Insulin Receptor Antibody, clone 21G12 is an antibody against phospho-Insulin Receptor (Tyr 1322) for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Insulin Receptor (Tyr 1322)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 21g12
Conjugate Unconjugated
Formulation Lyophilized in 2X PBS containing 0.09% sodium azide, PEG, and sucrose.
Gene Accession No. NM_000208.2; NM_001079817.1
Gene Symbols INSR, CD220, IR, HHF5
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Tyrosine 1322 on phosphorylated insulin receptor
Purification Method Subsequent thiophilic adsorption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Canine, Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.