Learn More
Phospho-IR (Tyr972) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-IR (Tyr972) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | 100μL of affinity purified rabbit polyclonal IgG in 50% storage buffer (PBS (without Mg2+ and Ca2+), pH 7.3 containing 1.0mg/mL BSA (IgG, protease free) and 0.05% sodium azide) and 50% glycerol. |
| Gene Symbols | INSR; CD220; HHF5; IR |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to the amino acid region encompassing the human phospho-IR (Tyr972 (960)) in the juxtamembrane domain. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.