Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-IRS-2 (Ser388) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071517MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-151-7MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-151-7MI Supplier MilliporeSigma Supplier No. 071517
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho-IRS-2 (Ser388) Antibody detects level of phospho-IRS-2 (Ser388) and has been published and validated for use in Western Blotting, Peptide Inhibition Assay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-IRS-2 (Ser388)
Applications Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols IRS2
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human IRS-2 phosphorylated Ser388.
Purification Method Affinity Purified
Quantity 200 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_003740
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.