Learn More
Phospho-IRS1 (Ser302 mouse/ Ser307 human) Mouse anti-Human, Mouse, Rat, Monkey, Bovine, Canine, Pig, Clone: 8.1.2, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-IRS1 (Ser302 mouse/ Ser307 human) |
| Applications | Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 8.1.2 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal antibody in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Accession No. | NP_005535.1 |
| Gene Symbols | HIRS-1 , IRS-1 |
| Host Species | Mouse |
| Immunogen | Synthetic linear peptide corresponding to amino acids at and around phosphorylated Ser302 of mouse IRS1. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.