Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-IRS1 (Ser612) mouse/ (Ser616) human Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 09448MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-448-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-448-MI Supplier MilliporeSigma Supplier No. 09448

Rabbit Polyclonal Antibody

Detect phospho-IRS1 (Ser612) mouse/ (Ser616) human using this Anti-phospho-IRS1 (Ser612) mouse/ (Ser616) human Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-IRS1 (Ser612) mouse/ (Ser616) human
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified rabbit polyclonal IgG in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH7.4, + 50% Glycerol.
Gene Symbols IRS1; IRS-1; HIRS-1
Host Species Rabbit
Immunogen peptide corresponding to amino acids encompassing and including phosphorylated Ser612 in mouse and Ser616 in human of IRS1
Purification Method Affinity Purfied
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_005544 (Human)
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.