Learn More
Phospho-ITCH (pT222) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-ITCH (pT222) |
| Applications | Dot Blot, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in Tris-citrate/phosphate buffer, pH 7-8, containing 0.1% sodium azide. |
| Gene Symbols | ITCH; NAPP1; dJ468O1.1; Itch; AIF4; AIP4 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to a portion of human ITCH encompassing the phosphorylated Threonine at position 222 (PPPpTPRRP). |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.