Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-JAK2 (Tyr317) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 071318MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-131-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-131-8MI Supplier MilliporeSigma Supplier No. 071318

Rabbit Polyclonal Antibody

Detect phospho-JAK2 (Tyr317) using this Anti-phospho-JAK2 (Tyr317) Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-JAK2 (Tyr317)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit serum with 0.05% sodium azide.
Gene Symbols EC 2.7.10.2; JAK-2; JTK10 ; OTTHUMP00000021024; OTTHUMP00000043260
Host Species Rabbit
Immunogen KLH-conjugated recombinant protein selected from residues corresponding to Tyr317 of JAK2.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_004963.1
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.