Learn More
Phospho-KDR/FLK-1/VEGFR2 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho KDR/FLK-1/VEGFR2 (Tyr1059) |
| Applications | Inhibition Assays, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Symbols | KDR; FLK1; VEGFR2 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human KDR/FLK-1/VEGFR2 phosphorylated at Tyr1059. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.