Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Lamellipodin/RAPH1 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABT279MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT279MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT279MI Supplier MilliporeSigma Supplier No. ABT279

Rabbit Polyclonal Antibody

This Anti-phospho-Lamellipodin/RAPH1 Antibody (Tyr426) is validated for use in Western Blotting and Peptide Inhibition Assay and Immunocytochemistry for the detection of phospho-Lamellipodin/RAPH1 (Tyr426).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-Lamellipodin/RAPH1 (Tyr426)
Applications Immunocytochemistry, Inhibition Assays, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit Polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols RAPH1; PREL2; RMO1; ALS2CR18; ALS2CR9; KIAA1681; LPD
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the PH domain of Human Lamellipodin/RAPH1 phosphorylated at Tyr426.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_998754
Target Species Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.