Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-LAT (Tyr191) Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07278
Description
This Anti-phospho-LAT (Tyr191) Antibody is validated for use in western blotting for the detection of phospho-LAT (Tyr191).
Specifications
phospho-LAT (Tyr191) | |
Polyclonal | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
Rabbit | |
Protein A chromatography | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
Unconjugated | |
LAT; LAT1; pp36; p36-38 | |
peptide (SREpYVNVSPEQQ-C) corresponding to amino acids 188-195 of human LAT and amino acids 192-203 of mouse LAT | |
200 μg | |
NM_014387 | |
−20°C for 24 months | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction