Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-LCK (Tyr505), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA126703
Description
PA1-26703 detects Lck (phospho Y505) from human samples. PA1-26703 is expected to cross react with chicken (92% conserved) and mouse (100% conserved) due to sequence homology.PA1-26703 has been successfully used in Western blot procedures.The PA1-26703 immunogen is a synthetic phosphopeptide corresponding to a portion of human Lck containing tyrosine 505. The sequence is conserved in human and mouse.
Specifications
Phospho-LCK pTyr505 | |
Polyclonal | |
LCK | |
LCK | |
Synthetic phosphopeptide corresponding to a portion of human Lck containing tyrosine 505. The sequence is conserved in human and mouse. | |
50 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
P06239, P06240 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3932 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction