Learn More
Phospho-LKB1 (Ser307) Rabbit anti-Human, Mouse, Rat, Bovine, Horse, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | phospho-LKB1 (Ser307) |
| Applications | Dot Blot, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. |
| Gene Symbols | STK11; LKB1; PJS |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to amino acids including and surrounding phosphorylated Ser307 of human LKB1. |
| Purification Method | Affinity Purified |
| Quantity | 200 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.