Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MAP Kinase1/2 Mouse anti-Human, Mouse, Rat, Avian, Canine, Clone: 12D4, MilliporeSigma™

Catalog No. 05481MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-481-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-481-MI Supplier MilliporeSigma Supplier No. 05481
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect phospho-MAP Kinase1/2 using this Anti-phospho-MAP Kinase1/2 validated for use in western blot, immunoprecipitation, ELISA and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-MAP Kinase1/2
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 12D4
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1k lyophilized from 1 mL 2x PBS / 0.09 % Na-azide / PEG and Sucrose.
Reconstitute with 1 mL H2O (15 min, RT) for a final concentration of 100μg/mL.
Gene Accession No. NM_002745, NM_002746
Gene Symbols MAPK3, MGC20180, ERT2, HUMKER1A, p44-MAPK, PRKM3, ERK1, p44erk1, p44mapk, P44MAPK, HS44KDAP, p44-ERK1, P44ERK1, ERK-1
Host Species Mouse
Immunogen KLH-conjugated phosphopeptide containing the pTEpY motif of Erk1 and Erk2. Clone: 12D4
Purification Method Thiophilic adsoption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Avian, Canine, Human, Mouse, Rat
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.