Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MAP1B (Thr1265) Rabbit anti-Rat, Mouse, Polyclonal, MilliporeSigma™

Catalog No. ABN58MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN58MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN58MI Supplier MilliporeSigma Supplier No. ABN58
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-phospho MAP1B (Thr1265) Antibody is an antibody against phospho MAP1B (Thr1265) for use in western blot, immunohistochemistry and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho MAP1B (Thr1265)
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in PBS containing 0.05% sodium azide.
Gene Symbols Map1b; Mtap1b; Mtap5
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to mouse MAP1B phosphorylated at Thr1265.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.