Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MARCKS (Ser152/156) Rabbit anti-Rat, Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 071238MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-123-8MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-123-8MI Supplier MilliporeSigma Supplier No. 071238
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-phospho-MARCKS (Ser152/156) Antibody is validated for use in western blot, immunohistochemistry (paraffin) for the detection of phospho-MARCKS (Ser152/156).
TRUSTED_SUSTAINABILITY

Specifications

Antigen phospho-MARCKS (Ser152/156)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in 10mM Hepes, pH 7.5, with 150mM NaCl, 100μg/mL BSA and 50% glycerol.
Gene Symbols MARCKS; MACS; 80K-L; PRKCSL; FLJ14368; phosphomyristin; MRACKS; PKCSL; FLJ90045
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids surrounding the phosphoSer152/156 of rat MARCKS.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002356.5
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.