Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MBP Mouse anti-Vertebrates, Clone: P12, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05429
Description
Detect phospho-MBP using this Anti-phospho-MBP Antibody, clone P12 validated for use in EA and western blotting.
Specifications
phospho-MBP | |
Monoclonal | |
Unconjugated | |
NM_002385 | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
IgG |
Western Blot | |
P12 | |
Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. | |
MBP, MGC99675 | |
Synthetic peptide corresponding to the human myelin basic protein sequence phosphorylated at Thr98 and coupled to tuberculin | |
100 μg | |
Vertebrates | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction